Assay Details
| Gene Symbol: arl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005947.1 | NP_001005947.1 | 1 - 2 | 186 | 76 | |
| GenBank mRNA | BC083457.1 | - | 1 - 2 | 186 | 76 | |
| BC153432.1 | - | 1 - 2 | 186 | 76 | ||
More Information
Set Membership: |
|

arl2
Genomic Map