Assay Details
| Gene Symbol: sgf29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005986.1 | NP_001005986.1 | 4 - 5 | 385 | 70 | |
| XM_005163810.4 | XP_005163867.1 | 4 - 5 | 457 | 70 | ||
| XM_005163811.4 | XP_005163868.1 | 4 - 5 | 463 | 70 | ||
| GenBank mRNA | BC083421.1 | - | 4 - 5 | 385 | 70 | |
More Information
Set Membership: |
|

sgf29
Genomic Map