Assay Details
Gene Symbol: mrps16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006030.1 | NP_001006030.1 | 3 - 4 | 393 | 72 | |
GenBank mRNA | BC083390.1 | - | 3 - 4 | 393 | 72 | |
BC164936.1 | - | 1 - 2 | 309 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
