Assay Details
| Gene Symbol: atp6v0e1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006038.2 | NP_001006038.2 | 1 - 2 | 210 | 64 | |
| GenBank mRNA | BC083377.1 | - | 1 - 2 | 192 | 64 | |
| BC164229.1 | - | 139 | 64 | |||
More Information
Set Membership: |
|

atp6v0e1
Genomic Map