Assay Details
| Gene Symbol: cutc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006046.1 | NP_001006046.1 | 4 - 5 | 373 | 79 | |
| GenBank mRNA | BC083362.1 | - | 4 - 5 | 373 | 79 | |
More Information
Set Membership: |
|

cutc
Genomic Map