Assay Details
| Gene Symbol: hsd17b10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006098.1 | NP_001006098.1 | 3 - 4 | 432 | 120 | |
| GenBank mRNA | BC083219.1 | - | 3 - 4 | 432 | 120 | |
| BC165246.1 | - | 3 - 4 | 391 | 120 | ||
More Information
Set Membership: |
|

hsd17b10
Genomic Map