Assay Details
| Gene Symbol: acin1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005171275.1 | XP_005171332.1 | 16 - 17 | 5430 | 102 | |
| XM_005171276.1 | XP_005171333.1 | 16 - 17 | 5427 | 102 | ||
| GenBank mRNA | BC116536.1 | - | 10 - 11 | 1303 | 102 | |
| CN504472.1 | - | 1 - 2 | 319 | 102 | ||
| DT078076.1 | - | 4 - 5 | 584 | 102 | ||
| EE691442.1 | - | 1 - 2 | 127 | 102 | ||
More Information
Set Membership: |
|

acin1a
Genomic Map