Assay Details
| Gene Symbol: acin1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005171275.1 | XP_005171332.1 | 11 - 12 | 4578 | 144 | |
| XM_005171276.1 | XP_005171333.1 | 11 - 12 | 4575 | 144 | ||
| GenBank mRNA | BC116536.1 | - | 5 - 6 | 451 | 144 | |
| CT659964.2 | - | 4 - 5 | 451 | 144 | ||
| CT699504.1 | - | 6 - 7 | 648 | 144 | ||
| EB785743.1 | - | 2 - 3 | 261 | 144 | ||
More Information
Set Membership: |
|

acin1a
Genomic Map