Assay Details
| Gene Symbol: ptpn4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007200.1 | NP_001007201.1 | 13 - 14 | 1207 | 128 | |
| GenBank mRNA | BC162720.1 | - | 13 - 14 | 1221 | 128 | |
More Information
Set Membership: |
|

ptpn4a
Genomic Map