Assay Details
Gene Symbol: gpr34b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001128546.1 | NP_001122018.1 | 1 | 955 | 116 | |
GenBank mRNA | BC092867.1 | - | 1 | 952 | 116 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
