Assay Details
| Gene Symbol: btg3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007351.2 | NP_001007352.1 | 3 - 4 | 457 | 91 | |
| NM_001326706.1 | NP_001313635.1 | 4 - 5 | 533 | 91 | ||
| NM_001326707.1 | NP_001313636.1 | 3 - 4 | 474 | 91 | ||
| NM_001326708.1 | NP_001313637.1 | 3 - 4 | 457 | 91 | ||
| GenBank mRNA | BC085526.1 | - | 3 - 4 | 407 | 91 | |
| BC164876.1 | - | 2 - 3 | 344 | 91 | ||
| EH539756.1 | - | 3 - 4 | 436 | 91 | ||
| EH593294.1 | - | 4 - 5 | 490 | 91 | ||
More Information
Set Membership: |
|

btg3
Genomic Map