Assay Details
Gene Symbol: churc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007380.1 | NP_001007381.1 | 3 - 4 | 302 | 76 | |
XM_021468411.1 | XP_021324086.1 | 3 - 4 | 276 | 76 | ||
GenBank mRNA | BC085457.1 | - | 3 - 4 | 302 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
