Assay Details
| Gene Symbol: ttc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007382.1 | NP_001007383.1 | 3 - 4 | 554 | 100 | |
| GenBank mRNA | BC085453.1 | - | 3 - 4 | 554 | 100 | |
| BC164119.1 | - | 2 - 3 | 503 | 100 | ||
More Information
Set Membership: |
|

ttc1
Genomic Map