Assay Details
Gene Symbol: pus1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008603.2 | NP_001008603.2 | 4 - 5 | 716 | 74 | |
GenBank mRNA | BC086827.1 | - | 4 - 5 | 621 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
