Assay Details
Gene Symbol: rnf7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012498.2 | NP_001012516.1 | 2 - 3 | 230 | 63 | |
GenBank mRNA | BC090163.1 | - | 2 - 3 | 234 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
