Assay Details
Gene Symbol: ndufa9a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013459.2 | NP_001013477.2 | 5 - 6 | 616 | 70 | |
GenBank mRNA | BC068378.1 | - | 5 - 6 | 544 | 70 | |
BC091545.1 | - | 5 - 6 | 566 | 70 | ||
BC152139.1 | - | 5 - 6 | 566 | 70 | ||
BC153422.1 | - | 5 - 6 | 1016 | 70 | ||
BC164086.1 | - | 5 - 6 | 594 | 70 | ||
DV588298.1 | - | 5 - 6 | 600 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
