Assay Details
Gene Symbol: capn2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013501.2 | NP_001013519.1 | 12 - 13 | 1605 | 152 | |
GenBank mRNA | BC090435.1 | - | 12 - 13 | 1609 | 152 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
