Assay Details
Gene Symbol: mat2ab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014296.2 | NP_001014318.1 | 1 - 2 | 150 | 90 | |
XM_017356170.2 | XP_017211659.1 | 1 - 2 | 163 | 90 | ||
GenBank mRNA | BC164405.1 | - | 1 - 2 | 123 | 90 | |
JN848835.1 | - | 1 - 2 | 146 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
