Assay Details
| Gene Symbol: smpdl3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014338.1 | NP_001014360.1 | 6 - 7 | 1007 | 75 | |
| XM_005170176.4 | XP_005170233.1 | 6 - 7 | 1097 | 75 | ||
| XM_009294917.3 | XP_009293192.1 | 6 - 7 | 1104 | 75 | ||
| GenBank mRNA | BC091833.1 | - | 6 - 7 | 1007 | 75 | |
More Information
Set Membership: |
|

smpdl3a
Genomic Map