Assay Details
| Gene Symbol: phox2bb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001014818.1 | NP_001014818.1 | 2 - 3 | 655 | 63 | |
| GenBank mRNA | AY846871.1 | - | 2 - 3 | 655 | 63 | |
| BC162881.1 | - | 2 - 3 | 555 | 63 | ||
| BC162887.1 | - | 2 - 3 | 555 | 63 | ||
More Information
Set Membership: |
|

phox2bb
Genomic Map