Assay Details
Gene Symbol: lgals3bp.2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017658.1 | NP_001017658.1 | 4 - 5 | 710 | 85 | |
GenBank mRNA | BC093341.1 | - | 4 - 5 | 710 | 85 | |
BC164862.1 | - | 4 - 5 | 690 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
