Assay Details
| Gene Symbol: grem2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017704.1 | NP_001017704.1 | 1 - 2 | 116 | 80 | |
| XM_009306988.3 | XP_009305263.1 | 1 - 2 | 339 | 80 | ||
| GenBank mRNA | BC093266.1 | - | 1 - 2 | 116 | 80 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC093266.1, NM_001017704.1 transcripts.
Set Membership: |
|

grem2b
Genomic Map