Assay Details
| Gene Symbol: dhrs7b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017758.1 | NP_001017758.1 | 6 - 7 | 774 | 71 | |
| XM_021470629.1 | XP_021326304.1 | 6 - 7 | 832 | 71 | ||
| GenBank mRNA | BC093357.1 | - | 6 - 7 | 774 | 71 | |
More Information
Set Membership: |
|

dhrs7b
Genomic Map