Assay Details
| Gene Symbol: crlf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017817.2 | NP_001017817.2 | 8 - 9 | 1161 | 71 | |
| GenBank mRNA | AM233512.1 | - | 8 - 9 | 1158 | 71 | |
| BC090429.1 | - | 4 - 5 | 71 | |||
| BC092800.1 | - | 8 - 9 | 71 | |||
| BC155810.1 | - | 8 - 9 | 71 | |||
More Information
Set Membership: |
|

crlf3
Genomic Map