Assay Details
| Gene Symbol: capns1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017899.2 | NP_001017899.2 | 7 - 8 | 530 | 102 | |
| GenBank mRNA | BC092946.1 | - | 3 - 4 | 274 | 102 | |
| CV490399.1 | - | 7 - 8 | 461 | 102 | ||
More Information
Set Membership: |
|

capns1a
Genomic Map