Assay Details
| Gene Symbol: mettl2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017902.1 | NP_001017902.1 | 6 - 7 | 753 | 98 | |
| GenBank mRNA | BC092943.1 | - | 6 - 7 | 753 | 98 | |
| BC151872.1 | - | 5 - 6 | 98 | |||
| BC154164.1 | - | 6 - 7 | 757 | 98 | ||
More Information
Set Membership: |
|

mettl2a
Genomic Map