Assay Details
| Gene Symbol: capn2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001018227.1 | NP_001018177.1 | 18 - 19 | 1962 | 119 | |
| GenBank mRNA | BC076522.1 | - | 19 - 20 | 1936 | 119 | |
| BC095045.1 | - | 18 - 19 | 1962 | 119 | ||
| BC155147.1 | - | 18 - 19 | 1913 | 119 | ||
| BC165647.1 | - | 18 - 19 | 1924 | 119 | ||
| BC171423.1 | - | 18 - 19 | 1913 | 119 | ||
More Information
Set Membership: |
|

capn2b
Genomic Map