Assay Details
| Gene Symbol: capn2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001018227.1 | NP_001018177.1 | 2 - 3 | 378 | 129 | |
| GenBank mRNA | BC076522.1 | - | 3 - 4 | 352 | 129 | |
| BC095045.1 | - | 2 - 3 | 378 | 129 | ||
| BC155147.1 | - | 2 - 3 | 329 | 129 | ||
| BC165647.1 | - | 2 - 3 | 340 | 129 | ||
| BC171423.1 | - | 2 - 3 | 329 | 129 | ||
More Information
Set Membership: |
|

capn2b
Genomic Map