Assay Details
| Gene Symbol: capn2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001018227.1 | NP_001018177.1 | 8 - 9 | 1042 | 115 | |
| GenBank mRNA | BC076522.1 | - | 9 - 10 | 1016 | 115 | |
| BC095045.1 | - | 8 - 9 | 1042 | 115 | ||
| BC155147.1 | - | 8 - 9 | 993 | 115 | ||
| BC165647.1 | - | 8 - 9 | 1004 | 115 | ||
| BC171423.1 | - | 8 - 9 | 993 | 115 | ||
More Information
Set Membership: |
|

capn2b
Genomic Map