Assay Details
Gene Symbol: cybrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020500.1 | NP_001018336.1 | 3 - 4 | 594 | 72 | |
GenBank mRNA | BC095170.1 | - | 3 - 4 | 594 | 72 | |
BC153653.1 | - | 3 - 4 | 594 | 72 | ||
BC165119.1 | - | 3 - 4 | 582 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
