Assay Details
Gene Symbol: ctss2.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024409.2 | NP_001019580.2 | 7 - 8 | 958 | 131 | |
GenBank mRNA | BC095694.1 | - | 7 - 8 | 131 | ||
BC095788.1 | - | 7 - 8 | 949 | 131 | ||
BC107612.1 | - | 7 - 8 | 958 | 131 | ||
BI980010.1 | - | 1 - 2 | 287 | 131 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
