Assay Details
| Gene Symbol: brms1la | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030102.2 | NP_001025273.1 | 8 - 9 | 1100 | 69 | |
| XM_005158646.4 | XP_005158703.1 | 8 - 9 | 1066 | 69 | ||
| XM_009292866.3 | XP_009291141.1 | 8 - 9 | 1063 | 69 | ||
| GenBank mRNA | BC097187.1 | - | 8 - 9 | 1108 | 69 | |
More Information
Set Membership: |
|

brms1la
Genomic Map