Assay Details
| Gene Symbol: ehmt1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030131.2 | NP_001025302.1 | 21 - 22 | 2807 | 114 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ehmt1a| Gene Symbol: ehmt1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030131.2 | NP_001025302.1 | 21 - 22 | 2807 | 114 | |
Set Membership: |
|