Assay Details
| Gene Symbol: ehmt1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030131.2 | NP_001025302.1 | 4 - 5 | 628 | 61 | |
| XM_005171778.4 | XP_005171835.1 | 5 - 6 | 800 | 61 | ||
| XM_005171779.4 | XP_005171836.1 | 5 - 6 | 800 | 61 | ||
More Information
Set Membership: |
|

ehmt1a
Genomic Map