Assay Details
Gene Symbol: tmx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030159.1 | NP_001025330.1 | 5 - 6 | 530 | 66 | |
GenBank mRNA | BC098617.1 | - | 5 - 6 | 530 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
