Assay Details
| Gene Symbol: mrps22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098735.1 | NP_001092205.1 | 5 - 6 | 681 | 74 | |
| GenBank mRNA | BC142888.1 | - | 5 - 6 | 681 | 74 | |
| BC150427.1 | - | 5 - 6 | 825 | 74 | ||
More Information
Set Membership: |
|

mrps22
Genomic Map