Assay Details
Gene Symbol: aste1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099430.2 | NP_001092900.1 | 4 - 5 | 1740 | 74 | |
GenBank mRNA | BC163487.1 | - | 4 - 5 | 1734 | 74 | |
BC163493.1 | - | 4 - 5 | 1734 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
