Assay Details
Gene Symbol: ahsg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100029.1 | NP_001093499.1 | 6 - 7 | 933 | 71 | |
GenBank mRNA | BC150351.1 | - | 6 - 7 | 779 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
