Assay Details
Gene Symbol: tdo2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102616.2 | NP_001096086.1 | 7 - 8 | 750 | 130 | |
GenBank mRNA | BC150376.1 | - | 7 - 8 | 752 | 130 | |
BC151920.1 | - | 7 - 8 | 752 | 130 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
