Assay Details
Gene Symbol: cyb5d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102674.1 | NP_001096144.1 | 2 - 3 | 589 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
