Assay Details
| Gene Symbol: cyb561a3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105584.1 | NP_001099054.1 | 6 - 7 | 914 | 62 | |
| XM_005171773.4 | XP_005171830.1 | 6 - 7 | 1051 | 62 | ||
| GenBank mRNA | BC093310.1 | - | 5 - 6 | 830 | 62 | |
| BC153668.1 | - | 6 - 7 | 914 | 62 | ||
More Information
Set Membership: |
|

cyb561a3a
Genomic Map