Assay Details
| Gene Symbol: gamt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105595.1 | NP_001099065.1 | 2 - 3 | 355 | 61 | |
| GenBank mRNA | BC107607.1 | - | 2 - 3 | 402 | 61 | |
| BC153601.1 | - | 2 - 3 | 355 | 61 | ||
More Information
Set Membership: |
|

gamt
Genomic Map