Assay Details
Gene Symbol: anxa2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105600.1 | NP_001099070.1 | 3 - 4 | 243 | 94 | |
GenBank mRNA | BC153582.1 | - | 3 - 4 | 243 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
