Assay Details
Gene Symbol: anxa2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105600.1 | NP_001099070.1 | 8 - 9 | 694 | 75 | |
GenBank mRNA | BC153582.1 | - | 8 - 9 | 694 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
