Assay Details
| Gene Symbol: capns1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109706.1 | NP_001103176.1 | 10 - 11 | 796 | 126 | |
| XM_005157735.1 | XP_005157792.1 | 10 - 11 | 736 | 126 | ||
| GenBank mRNA | BC151952.1 | - | 10 - 11 | 796 | 126 | |
| BC162123.1 | - | 10 - 11 | 782 | 126 | ||
| BC162479.1 | - | 10 - 11 | 782 | 126 | ||
More Information
Set Membership: |
|

capns1b
Genomic Map