Assay Details
| Gene Symbol: vdac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001404.1 | NP_001001404.1 | 7 - 8 | 775 | 138 | |
| GenBank mRNA | AY398429.1 | - | 7 - 8 | 775 | 138 | |
| BC067647.1 | - | 7 - 8 | 737 | 138 | ||
More Information
Set Membership: |
|

vdac1
Genomic Map