Assay Details
| Gene Symbol: slc39a6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001591.1 | NP_001001591.1 | 5 - 6 | 1226 | 56 | |
| XM_005171365.4 | XP_005171422.1 | 5 - 6 | 1264 | 56 | ||
| GenBank mRNA | AB126260.1 | - | 5 - 6 | 1226 | 56 | |
More Information
Set Membership: |
|

slc39a6
Genomic Map