Assay Details
| Gene Symbol: sox3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001811.2 | NP_001001811.2 | 1 | 854 | 109 | |
| GenBank mRNA | AB242330.1 | - | 1 | 802 | 109 | |
| AY316135.1 | - | 1 | 800 | 109 | ||
| BC092845.1 | - | 1 | 854 | 109 | ||
| BC155138.1 | - | 1 | 826 | 109 | ||
More Information
Set Membership: |
|

sox3
Genomic Map