Assay Details
| Gene Symbol: igf2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001815.1 | NP_001001815.1 | 3 - 4 | 489 | 99 | |
| GenBank mRNA | AF250289.1 | - | 3 - 4 | 489 | 99 | |
More Information
Set Membership: |
|

igf2b
Genomic Map