Assay Details
Gene Symbol: lgals9l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001817.1 | NP_001001817.1 | 3 - 4 | 715 | 95 | |
GenBank mRNA | AY394929.1 | - | 3 - 4 | 715 | 95 | |
BC076011.1 | - | 3 - 4 | 390 | 95 | ||
BC095566.1 | - | 3 - 4 | 716 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
